Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrtm3
Entrez Gene ID
100759319
Gene Name
leucine rich repeat transmembrane neuronal 3
Gene Aliases
I79_007362
Chromosome Location
Chr.Un|NW_003613826: 1039579 - 1196006 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501001.2 XP_003501049.1 1 - 2 1533 74

View Full Product Details