Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rap2c
Entrez Gene ID
100761576
Gene Name
RAP2C, member of RAS oncogene family
Gene Aliases
I79_013378
Chromosome Location
Chr.Un|NW_003614212: 225370 - 238923 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506103.2 XP_003506151.1 2 - 3 615 100
GenBank mRNA JI867583.1 - 2 - 3 524 100

View Full Product Details