Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hrh3
Entrez Gene ID
100764853
Gene Name
histamine receptor H3
Gene Aliases
I79_009854
Chromosome Location
Chr.Un|NW_003613943: 33339 - 35786 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643786.1 XP_007641976.1 2 - 3 615 83

View Full Product Details