Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tomm6
Entrez Gene ID
100770746
Gene Name
translocase of outer mitochondrial membrane 6 homolog (yeast)
Gene Aliases
I79_017054
Chromosome Location
Chr.Un|NW_003614620: 436237 - 437626 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508992.2 XP_003509040.1 1 - 2 311 62
GenBank mRNA JI865671.1 - 1 - 2 204 62

View Full Product Details