Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dcps
Entrez Gene ID
100761162
Gene Name
decapping enzyme, scavenger
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613908: 1073574 - 1118149 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643315.1 XP_007641505.1 3 - 4 596 75
GenBank mRNA JI865328.1 - 3 - 4 566 75

View Full Product Details