Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Akap17a
Entrez Gene ID
100769666
Gene Name
A kinase (PRKA) anchor protein 17A
Gene Aliases
I79_006207
Chromosome Location
Chr.Un|NW_003613768: 680847 - 711741 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500032.2 XP_003500080.1 4 - 5 1051 85

View Full Product Details