Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Serpina9
Entrez Gene ID
100761678
Gene Name
serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 9
Gene Aliases
I79_017527
Chromosome Location
Chr.Un|NW_003614700: 499272 - 509733 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509369.1 XP_003509417.1 3 - 4 1064 67

View Full Product Details