Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sdhaf2
Entrez Gene ID
100772110
Gene Name
succinate dehydrogenase complex assembly factor 2
Gene Aliases
I79_022213
Chromosome Location
Chr.Un|NW_003615831: 135722 - 158285 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513278.2 XP_003513326.2 2 - 3 393 114
GenBank mRNA JP040321.1 - 2 - 3 393 114

View Full Product Details