Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hesx1
Entrez Gene ID
100770203
Gene Name
HESX homeobox 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613611: 605736 - 607185 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496104.1 XP_003496152.1 3 - 4 457 82

View Full Product Details