Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wfikkn2
Entrez Gene ID
100766981
Gene Name
WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613846: 1339447 - 1343646 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501379.1 XP_003501427.1 1 - 2 187 60

View Full Product Details