Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pmch
Entrez Gene ID
100770992
Gene Name
pro-melanin-concentrating hormone
Gene Aliases
Pro-MCH
Chromosome Location
Chr.Un|NW_003613751: 985372 - 986267 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007640422.1 XP_007638612.1 1 - 2 314 89

View Full Product Details