Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cldnd2
Entrez Gene ID
100752776
Gene Name
claudin domain containing 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615616: 126954 - 128604 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512842.1 XP_003512890.1 1 - 2 170 73

View Full Product Details