Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ppcs
Entrez Gene ID
100768962
Gene Name
phosphopantothenoylcysteine synthetase
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615864: 175921 - 179610 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513331.2 XP_003513379.1 1 - 2 819 55

View Full Product Details