Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nfe2l3
Entrez Gene ID
103162263
Gene Name
nuclear factor, erythroid 2-like 3
Gene Aliases
I79_004412
Chromosome Location
Chr.Un|NW_003613695: 676105 - 872477 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007629800.1 XP_007627990.1 4 - 5 521 91

View Full Product Details