Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dlx6
Entrez Gene ID
100770813
Gene Name
distal-less homeobox 6
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613851: 1212063 - 1218392 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501473.2 XP_003501521.1 1 - 2 366 82

View Full Product Details