Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Grp
Entrez Gene ID
100769410
Gene Name
gastrin-releasing peptide
Gene Aliases
I79_013760
Chromosome Location
Chr.Un|NW_003614238: 37154 - 52241 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506395.2 XP_003506443.2 3 - 4 481 77

View Full Product Details