Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankrd23
Entrez Gene ID
100769457
Gene Name
ankyrin repeat domain 23
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613687: 967111 - 976281 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007625771.1 XP_007623961.1 5 - 6 556 104

View Full Product Details