Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Apoa1bp
Entrez Gene ID
100762326
Gene Name
apolipoprotein A-I binding protein
Gene Aliases
I79_009597
Chromosome Location
Chr.Un|NW_003613923: 921909 - 924194 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643549.1 XP_007641739.1 5 - 6 943 88
GenBank mRNA JI871085.1 - 4 - 5 487 88

View Full Product Details