Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sf3b5
Entrez Gene ID
100756721
Gene Name
splicing factor 3b, subunit 5, 10kDa
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613761: 1213873 - 1214557 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499932.2 XP_003499980.1 1 - 1 321 62

View Full Product Details