Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Creg2
Entrez Gene ID
100759597
Gene Name
cellular repressor of E1A-stimulated genes 2
Gene Aliases
I79_005956
Chromosome Location
Chr.Un|NW_003613754: 2122097 - 2155311 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499805.1 XP_003499853.1 1 - 2 436 57

View Full Product Details