Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Npffr1
Entrez Gene ID
100768907
Gene Name
neuropeptide FF receptor 1
Gene Aliases
I79_006046
Chromosome Location
Chr.Un|NW_003613759: 987614 - 1002778 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499903.1 XP_003499951.1 5 - 6 870 101

View Full Product Details