Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chadl
Entrez Gene ID
100763136
Gene Name
chondroadherin-like
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614859: 127391 - 134715 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007650945.1 XP_007649135.1 5 - 6 1721 76

View Full Product Details