Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrp3
Entrez Gene ID
100769612
Gene Name
low density lipoprotein receptor-related protein 3
Gene Aliases
I79_020064
Chromosome Location
Chr.Un|NW_003615142: 20886 - 24851 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007652080.1 XP_007650270.1 4 - 5 1105 75

View Full Product Details