Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim42
Entrez Gene ID
100759091
Gene Name
tripartite motif containing 42
Gene Aliases
I79_000740
Chromosome Location
Chr.Un|NW_003613594: 1485065 - 1508540 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495484.1 XP_003495532.1 1 - 2 339 93

View Full Product Details