Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bcmo1
Entrez Gene ID
100760350
Gene Name
beta-carotene 15,15'-monooxygenase 1
Gene Aliases
I79_000299
Chromosome Location
Chr.Un|NW_003613583: 6426241 - 6483524 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007609082.1 XP_007607272.1 5 - 6 506 92

View Full Product Details