Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Isl2
Entrez Gene ID
100764443
Gene Name
ISL LIM homeobox 2
Gene Aliases
I79_003585
Chromosome Location
Chr.Un|NW_003613669: 1294397 - 1298788 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497929.1 XP_003497977.1 5 - 6 574 90

View Full Product Details