Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hey2
Entrez Gene ID
100755611
Gene Name
hes-related family bHLH transcription factor with YRPW motif 2
Gene Aliases
I79_020832
Chromosome Location
Chr.Un|NW_003615372: 78524 - 87749 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512148.1 XP_003512196.1 5 - 6 541 94

View Full Product Details