Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mettl21b
Entrez Gene ID
100761001
Gene Name
methyltransferase like 21B
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614393: 700059 - 708874 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507515.2 XP_003507563.1 1 - 2 499 109

View Full Product Details