Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clns1a
Entrez Gene ID
100768284
Gene Name
chloride channel, nucleotide-sensitive, 1A
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614874: 369349 - 389318 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510396.2 XP_003510444.1 5 - 6 748 68
GenBank mRNA JI873405.1 - 5 - 6 738 68
JP064031.1 - 5 - 6 738 68

View Full Product Details