Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Il19
Entrez Gene ID
100774163
Gene Name
interleukin 19
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613661: 2985608 - 3007393 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007616651.1 XP_007614841.1 1 - 2 332 83
XM_007616658.1 XP_007614848.1 3 - 4 314 83
XM_007616666.1 XP_007614856.1 1 - 2 334 83

View Full Product Details