Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rdh5
Entrez Gene ID
100772460
Gene Name
retinol dehydrogenase 5 (11-cis/9-cis)
Gene Aliases
I79_013524
Chromosome Location
Chr.Un|NW_003614217: 931789 - 939737 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007646722.1 XP_007644912.1 2 - 3 1904 81

View Full Product Details