Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mrps14
Entrez Gene ID
100752801
Gene Name
mitochondrial ribosomal protein S14
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613658: 2307134 - 2312154 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497673.2 XP_003497721.1 1 - 2 256 76
GenBank mRNA JI889736.1 - 1 - 2 100 76
JP056682.1 - 1 - 2 100 76

View Full Product Details