Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hamp
Entrez Gene ID
100757672
Gene Name
hepcidin antimicrobial peptide
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003617758: 37907 - 39635 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003515310.2 XP_003515358.1 2 - 3 154 99

View Full Product Details