Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hipk4
Entrez Gene ID
100763406
Gene Name
homeodomain interacting protein kinase 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614129: 522515 - 530695 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505239.1 XP_003505287.1 2 - 3 823 75

View Full Product Details