Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plin4
Entrez Gene ID
100768515
Gene Name
perilipin 4
Gene Aliases
I79_008172
Chromosome Location
Chr.Un|NW_003613861: 296752 - 306104 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007642409.1 XP_007640599.1 3 - 4 317 81

View Full Product Details