Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
St13
Entrez Gene ID
100763998
Gene Name
suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein)
Gene Aliases
I79_018582
Chromosome Location
Chr.Un|NW_003614859: 416128 - 456030 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510309.2 XP_003510357.1 2 - 3 103 93

View Full Product Details