Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmed8
Entrez Gene ID
100764153
Gene Name
transmembrane emp24 protein transport domain containing 8
Gene Aliases
I79_005792
Chromosome Location
Chr.Un|NW_003613749: 1858075 - 1891156 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007640398.1 XP_007638588.1 5 - 6 742 97

View Full Product Details