Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lair1
Entrez Gene ID
100773832
Gene Name
leukocyte-associated immunoglobulin-like receptor 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615066: 86177 - 97806 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007651780.1 XP_007649970.1 3 - 4 482 144

View Full Product Details