Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccl21
Entrez Gene ID
100770444
Gene Name
chemokine (C-C motif) ligand 21
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614108: 982219 - 982878 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504977.1 XP_003505025.1 2 - 3 184 101

View Full Product Details