Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aqp5
Entrez Gene ID
100773918
Gene Name
aquaporin 5
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614438: 53036 - 55954 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507837.1 XP_003507885.1 1 - 2 364 70

View Full Product Details