Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Irs1
Entrez Gene ID
100755710
Gene Name
insulin receptor substrate 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615619: 163634 - 214386 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512849.2 XP_003512897.1 1 - 2 4260 83

View Full Product Details