Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ogfrl1
Entrez Gene ID
100757269
Gene Name
opioid growth factor receptor-like 1
Gene Aliases
I79_001301
Chromosome Location
Chr.Un|NW_003613608: 3387360 - 3424800 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007645866.1 XP_007644056.1 5 - 6 552 70

View Full Product Details