Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syt14
Entrez Gene ID
100765116
Gene Name
synaptotagmin XIV
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613696: 801714 - 861767 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007629918.1 XP_007628108.1 2 - 3 331 61
XM_007629924.1 XP_007628114.1 2 - 3 331 61

View Full Product Details