Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nefl
Entrez Gene ID
100774151
Gene Name
neurofilament, light polypeptide
Gene Aliases
I79_001527
Chromosome Location
Chr.Un|NW_003613613: 3896329 - 3899668 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496188.2 XP_003496236.1 2 - 3 1173 66

View Full Product Details