Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Meox2
Entrez Gene ID
100753046
Gene Name
mesenchyme homeobox 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614118: 983036 - 1046315 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505068.2 XP_003505116.1 2 - 3 896 85

View Full Product Details