Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hoxa11
Entrez Gene ID
100763010
Gene Name
homeobox A11
Gene Aliases
I79_008459
Chromosome Location
Chr.Un|NW_003613870: 95930 - 98217 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007642619.1 XP_007640809.1 2 - 3 273 100

View Full Product Details