Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Icoslg
Entrez Gene ID
100768618
Gene Name
inducible T-cell co-stimulator ligand
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613799: 1751746 - 1762210 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500605.1 XP_003500653.1 6 - 7 1498 89

View Full Product Details