Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC103163503
Entrez Gene ID
103163503
Gene Name
putative killer cell immunoglobulin-like receptor like protein KIR3DP1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615066: 60895 - 72275 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007651778.1 XP_007649968.1 1 - 2 73 75

View Full Product Details