Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Efhd2
Entrez Gene ID
100752661
Gene Name
EF-hand domain family, member D2
Gene Aliases
I79_020086
Chromosome Location
Chr.Un|NW_003615153: 118296 - 134872 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511544.2 XP_003511592.1 1 - 2 75 84
GenBank mRNA JI895974.1 - 1 - 2 48 84

View Full Product Details