Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aicda
Entrez Gene ID
100689250
Gene Name
activation-induced cytidine deaminase
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614669: 511273 - 519417 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007649816.1 XP_007648006.1 2 - 3 420 120

View Full Product Details